Order Entry
Puerto Rico
ContactUsLinkComponent
Human/Mouse Recombinant Activin A, ACF
Human/Mouse Recombinant Activin A, ACF
Catalog # MSPP-78132-1
CAS Number:  
undefined
Human/Mouse Recombinant Activin A, ACF
Catalog # MSPP-78132-1
Supplier Number:  78132.1
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
    Mouse
  • Size:
    100 µg
  • Protein/Peptide Name:
    Activin A, ACF
  • Purity:
    ≥95%
  • Cat. No.:
    MSPP-78132-1

Specifications

About this item

Activin A is a member of the transforming growth factor beta (TGF-β) family of proteins produced by many cell types throughout development (Gurdon et al.). It is a disulfide-linked homodimer (two beta-A chains) that binds to heteromeric complexes of a type I (Act RI-A and Act RI-B) and a type II (Act RII-A and Act RII-B) serine-threonine kinase receptor (Attisano et al.). Activins primarily signal through SMAD2/3 proteins to regulate a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (McDowell et al.). Activin A maintains the undifferentiated state of human embryonic stem cells (James et al.; Xiao et al.) and also facilitates differentiation of human embryonic stem cells into definitive endoderm (D’Amour et al.). This product is animal component-free.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').