Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant EPO (CHO-expressed)
Human Recombinant EPO (CHO-expressed)
Catalog # MSPP-78007
CAS Number:  
undefined
Human Recombinant EPO (CHO-expressed)
Catalog # MSPP-78007
Supplier Number:  78007
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Size:
    50 µg
  • Protein Synonyms:
    EP|Epoetin|Erythropoietin
  • Protein/Peptide Name:
    EPO
  • Purity:
    ≥95%
  • Cat. No.:
    MSPP-78007

Specifications

About this item

The principal physiological regulator of erythropoiesis, Erythropoietin (EPO) is a glycoprotein growth factor that is produced primarily in the kidney in response to hypoxia or anemia. EPO promotes erythropoiesis by binding to a homodimeric cell surface receptor that activates JAK2/STAT5, PI3K/AKT, and MAPK pathways, and stimulates the proliferation and differentiation of erythroid progenitor cells (Jelkmann. Transfus Med Hemother, 2013; Kuhrt and Wojchowski. Blood, 2015).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').