Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IFN-alpha 2A
Human Recombinant IFN-alpha 2A
Catalog # MSPP-78076
CAS Number:  
undefined
Human Recombinant IFN-alpha 2A
Catalog # MSPP-78076
Supplier Number:  78076
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Size:
    20 µg
  • Protein/Peptide Name:
    IFN-alpha 2A
  • Purity:
    ≥95%
  • Cat. No.:
    MSPP-78076

Specifications

About this item

Interferon-alpha (IFN-α) is a type I interferon, produced by virus-infected cells, and is released as a soluble factor to initiate antiviral responses (Isaacs and Lindenmann). IFN-α2 is the most potent IFN-α used in fundamental research and in most clinical applications. The best-known IFN-α2 subvariants, 2A and 2B, differ by only one or two amino acids at positions 23 and/or 34 of the mature protein (von Gabain et al.). Type I IFNs exert potent antitumor activity by increasing the cytotoxic activity of NK and T cells, as well as by inhibiting the proliferation of cancer cells (Paul et al.). Additionally, it has been shown that proinflammatory IFN-α modulates the function of B cells in patients with systemic lupus erythematosus (Chang et al.), and pegylated forms of IFN-alpha 2A and 2B have implications in the treatment of hepatitis C (Foster et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').