Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Human Recombinant BDNF

Human Recombinant BDNF

Marketsource
Human Recombinant BDNF
Marketsource
Certificates
Human Recombinant BDNF

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Brain-derived neurotrophic factor (BDNF), like nerve growth factor (NGF), neurotrophin-3 (NT-3), and neurotrophin-4 (NT-4), is a member of the NGF family of neurotrophins, which are required for the differentiation and survival of specific neuronal subpopulations in both the central and the peripheral nervous systems (Minichiello and Klein; Minichiello et al.). BDNF binds with high affinity to the tropomyosin receptor kinase B (TrkB), and activates AKT and ERK pathways (Mattson et al.). It is expressed in the hippocampus, cortex, and synapses of the basal forebrain. BDNF acts as a survival factor for human embryonic stem cells when plated on either feeder cells or Corning® Matrigel® (Pyle et al.). BDNF regulates synaptic transmission and plasticity at adult synapses in the central nervous system, and contributes to adaptive neuronal responses including long-term potentiation, long-term depression, certain forms of short-term synaptic plasticity, and homeostatic regulation of neuronal excitability (Reichardt). It also has a role in neurogenesis by promoting survival and growth of dorsal root ganglion cells, and hippocampal and cortical neurons (Binder and Scharfman). BDNF, together with glial cell line-derived neurotrophic factor (GDNF) and other supplements, is commonly used to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into neurons (Brafman).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • MSPP-78005
  • MSPP-78005-1
  • Protein/Peptide Name:
  • BDNF
  • BDNF
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 10 µg
    Log in for availability
    Log in for availability
    Each
    Each - $321.54
    $321.54
    Each
       $321.54
    Minimum order quantity is 1

    Supplier Number: 78005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MSPP-78005
    Protein/Peptide Name
    BDNF
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 100 µg
    Log in for availability
    Log in for availability
    Each
    Each - $1,734.85
    $1,734.85
    Each
       $1,734.85
    Minimum order quantity is 1

    Supplier Number: 78005.1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MSPP-78005-1
    Protein/Peptide Name
    BDNF
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.