Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IGF-I
Human Recombinant IGF-I
Certificates
undefined
Human Recombinant IGF-I

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Protein/Peptide Name:
    IGF-I
  • Purity:
    ≥95%

Specifications