Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IFN-gamma
Human Recombinant IFN-gamma
Catalog # MSPP-780201
CAS Number:  
undefined
Human Recombinant IFN-gamma
Catalog # MSPP-780201
Supplier Number:  78020.1
CAS Number:  

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein/Peptide Name:
    IFN-gamma
  • Purity:
    ≥95%
  • Cat. No.:
    MSPP-780201

Specifications

About this item

Interferon-gamma (IFN-γ), also known as type II interferon, is produced by T and NK cells, and in smaller amounts by dendritic cells and macrophages. IFN-γ is controlled by cytokines such as IL-12 and IL-18 secreted in response to infection (Schroder et al.). IFN-γ binds to a receptor complex and initiates signal transduction via the JAK/STAT pathway; this culminates in the transcription and activation of many genes that control a diverse array of immunological functions (de Weerd and Nguyen; Krause et al.). IFN-γ stimulates the antimicrobial and anti-tumor activity of macrophages, NK cells, and neutrophils (Billiau and Matthys) by promoting the activation of microbial effector functions such as production of reactive oxygen species, nitric oxide, and complement (Schroder et al.). IFN-γ enhances MHC class I and II expression in dendritic cells and mononuclear phagocytes, as well as the production of IL-12 by dendritic cells. In B cells, IFN-γ stimulates survival and growth in both mouse and human cells, and redirects B cells from proliferation towards differentiation. IFN-γ favors the development of Th1 vs Th2 cells and stimulates monocyte differentiation and function (Schroder et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').