Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:HRP
- Format:Pre-coated
- Primary antibody reactivity:Pig
- Target protein:Albumin
- Description:Porcine albumin ELISA
- Size:1×96 wells
- Sample Type:Plasma, serum, urine
- Detection Method:Colorimetric
- Time to Results:1 h, 40 min
- Detection Range:12.5 - 400 ng/ml
- Regulatory Status:RUO
- Cat. No.:102853-498
- Supplier no.:41-ALBPO-E01
The procine albumin ELISA is a highly sensitive two-site enzyme linked immunoassay for measuring albumin in porcine serum, tissue extracts and other biological fluids.
Albumin is a polyfunctional protein contributing to homeostasis through mechanisms of hemodynamics, transport and nutrition. Albumin is found both intra and extravascularly in all mammals and many lower vertebrates. It is a molecule of about 67,000 daltons, synthesized by the liver. Normally only very trace amounts of albumin escape reabsorption by kidney glomeruli and is excreted into the urine. Many occult diseases can cause kidney damage which may result in excessive amounts of serum proteins, including albumin, to be excreted by the kidney and into the urine.