Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Conjugate ELISA:HRP
- Format:Pre-coated
- Primary antibody reactivity:Chicken
- Target protein:Alpha 1-acid glycoprotein
- Description:Chicken alpha 1-acid glycoprotein ELISA
- Size:1×96 wells
- Sample Type:Plasma, serum
- Detection Method:Colorimetric
- Time to Results:1 h, 15 min
- Detection Range:6.25 - 200 ng/ml
- Sensitivity:6.25 ng/ml
- Regulatory Status:RUO
- Cat. No.:102672-788
- Supplier no.:41-AGPCH-E01
Specifications
About this item
The chicken alpha 1-acid glycoprotein ELISA is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring AGP in the biological fluids of chickens.
AGP is a 52 kDa serine protease inhibitor (serpin) in blood, which protects tissue from enzymes from inflammatory cells, especially elastase. In certain acute phase inflammatory reactions, AGP is elevated in order to limit the damage caused by activated neutrophil granulocytes and their enzyme elastase. Disorders of AGP include AGP deficiency, a hereditary disorder that can lead to severe tissue breakdown during inflammation. This may result in pulmonary emphysema and liver cirrhosis, in severe cases. Genetic variants of AGP do occur.