Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Alpha 1-Antitrypsin ELISA
Mouse Alpha 1-Antitrypsin ELISA
Catalog # 101720-928
Supplier:  Alpco Diagnostics
CAS Number:  
Mouse Alpha 1-Antitrypsin ELISA
Catalog # 101720-928
Supplier:  Alpco Diagnostics
Supplier Number:  41-A1AMS-E01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Alpha 1-antitrypsin
  • Description:
    Mouse Alpha 1-antitrypsin ELISA 
  • Size:
    1×96 wells
  • Sample Type:
    Serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h, 5 min
  • Detection Range:
    6.25 - 200 ng/ml
  • Sensitivity:
    6.25 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    101720-928
  • Supplier no.:
    41-A1AMS-E01

Specifications

About this item

The mouse alpha 1-antitrypsin ELISA is for the quantitative determination of a-1-antitrypsin in mouse serum.

Alpha 1-antitrypsin is a 52 kDa serine protease inhibitor (serpin) in blood, which protects tissue from enzymes from inflammatory cells, especially elastase. Research demonstrates that in certain acute phase inflammatory reactions, alpha 1-antitrypsin is elevated in order to limit the damage caused by activated neutrophil granulocytes and their enzyme elastase. Disorders of alpha 1-antitrypsin include alpha 1- antitrypsin deficiency, a hereditary disorder that can lead to severe tissue breakdown during inflammation. This may result in pulmonary emphysema and liver cirrhosis, in severe cases. Genetic variants of alpha 1- antitrypsin do occur.

Caution: For research use only. Not for use in diagnostic procedures.