Order Entry
Puerto Rico
ContactUsLinkComponent
Bi-CAT ELISA (Epinephrine & Norepinephrine)
Bi-CAT ELISA (Epinephrine & Norepinephrine)
Catalog # 101721-322
Supplier:  Alpco Diagnostics
CAS Number:  
Bi-CAT ELISA (Epinephrine & Norepinephrine)
Catalog # 101721-322
Supplier:  Alpco Diagnostics
Supplier Number:  17-BCTHU-E02.1
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Competitive
  • Conjugate ELISA:
    Peroxidase
  • Format:
    Pre-coated
  • Host:
    Goat
  • Primary antibody reactivity:
    Human
  • Target protein:
    Epinephrine and norepinephrine
  • Description:
    Bi-CAT (epinephrine and norepinephrine) ELISA
  • Size:
    2×96 wells
  • Sample Type:
    Plasma, urine
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h
  • Regulatory Status:
    RUO
  • Cat. No.:
    101721-322
  • Supplier no.:
    17-BCTHU-E02.1

Specifications

About this item

The Bi-CAT ELISA (Epinephrine and Norepinephrine ELISA) is an enzyme Immunoassay for the quantitative determination of adrenaline (epinephrine) and noradrenaline (norepinephrine) in plasma and urine.

Adrenaline (epinephrine) and noradrenaline (norepinephrine) are extracted by using a cis-diol-specific affinity gel, acylated and then derivatized enzymatically. The competitive ELISA kit uses the microtiter plate format. The antigen is bound to the solid phase of the microtiter plate. The derivatized standards, controls and samples and the solid phase bound analytes compete for a fixed number of antiserum binding sites. After the system is in equilibrium, free antigen and free antigen-antiserum complexes are removed by washing. The antibody bound to the solid phase is detected by an anti-rabbit IgG-peroxidase conjugate using TMB as a substrate. The reaction is monitored at 450 nm.

Caution: For research use only. Not for use in diagnostic procedures.