Order Entry
Puerto Rico
ContactUsLinkComponent
Rat High Range Insulin ELISA
Rat High Range Insulin ELISA
Catalog # 101720-332
Supplier:  Alpco Diagnostics
CAS Number:  
Rat High Range Insulin ELISA
Catalog # 101720-332
Supplier:  Alpco Diagnostics
Supplier Number:  80-INSRTH-E01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Insulin
  • Description:
    Rat high range insulin ELISA
  • Size:
    1×96 wells
  • Sample Type:
    Plasma, serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h
  • Detection Range:
    3 - 150 ng/ml
  • Sample Volume:
    5 µl
  • Sensitivity:
    0.52 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    101720-332
  • Supplier no.:
    80-INSRTH-E01

Specifications

About this item

The rat high range insulin ELISA quantifies the concentration of insulin protein products in rat I and rat II proinsulin genes with as little as 5 µl of serum or plasma sample.

  • Only 5 µl of sample is sufficient for the assay
  • Superior sensitivity of 0.52 ng/ml
  • Dynamic range of 3 to 150 ng/ml

Utilizing a dual-monoclonal antibody sandwich ELISA format, bi-level control set, and 96-well microplate comprising removable strips, the rat high range insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if an entire plate is not needed at one time.

Caution: For research use only. Not for use in diagnostic procedures.