ContactUsLinkComponent
SP Bel-Art Stainless Steel Pipettor Stand, Bel-Art Products, a part of SP
SP Bel-Art Stainless Steel Pipettor Stand, Bel-Art Products, a part of SP
Catalog # 53576-220
undefined
SP Bel-Art Stainless Steel Pipettor Stand, Bel-Art Products, a part of SP
Catalog # 53576-220
Supplier Number:  H18962-0006
Specifications
  • Dimensions:
    22.9W x 14.6L x 19.7H cm (9 x 53/4 x 73/4")
  • Cat. No.:
    53576-220
  • Supplier no.:
    189620006
Specifications
About this item
Made of stainless steel and without any plastic coating, this universal rack holds all types of pipettors, including the Gilson Pipetman®.
  • Stainless steel construction
  • Free-standing on benchtop
  • Compatible with all types of pipettors
It has five positions, a sloping back, and open front notches to hold pipettors in place. Suitable for free-standing use on a benchtop.