ContactUsLinkComponent
Filter Weighing Kit for XPR/XSR Analytical Balances
Filter Weighing Kit for XPR/XSR Analytical Balances
Catalog # 76310-512
Supplier:  Mettler Toledo
undefined
Filter Weighing Kit for XPR/XSR Analytical Balances
Catalog # 76310-512
Supplier:  Mettler Toledo
Supplier Number:  30460857
Specifications
  • Description:
    Filter weighing kit for XPR/XSR analytical balances
  • Cat. No.:
    76310-512
Specifications
About this item
Enables fast and easy weighing of filter samples up to 110 mm in diameter.
  • Special weighing cage allows precise positioning of filters
  • Three month warranty