Order Entry
Puerto Rico
ContactUsLinkComponent
Nalgene® Centrifuge Bottle with Screw Cap, Polycarbonate, Spherical Bottom, Thermo Scientific
Nalgene® Centrifuge Bottle with Screw Cap, Polycarbonate, Spherical Bottom, Thermo Scientific
  21020-120
undefined
Nalgene® Centrifuge Bottle with Screw Cap, Polycarbonate, Spherical Bottom, Thermo Scientific
  21020-120
 :  3123-0250

 

  • Ø:
    61.9 mm
  • Capacity:
    250 ml (8.5 oz.)
  • Description:
    Bottle with Cap
  • Length:
    145.8 mm with cap
  • Cat. No.:
    21020-120
  • Max. rcf:
    27500×g
  • Supplier no.:
    3123-0250

 

 

Transparent bottle is manufactured from polycarbonate and is leakproof at low speeds.

Note: When autoclaving, place cap on container without engaging threads. Must be used with spherical-bottom centrifuge bottle support 21020-142. Use sealing cap assembly 21009-901 for hazardous material or high speed. For proper performance, fill bottles to at least 80% of the total capacity.

Polypropylene closure. Autoclavable.