ContactUsLinkComponent
Synthware Heat Transfer Block for Round Bottom Flasks, Microscale, Kemtech America
Synthware Heat Transfer Block for Round Bottom Flasks, Microscale, Kemtech America
Catalog # 60004-230
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware Heat Transfer Block for Round Bottom Flasks, Microscale, Kemtech America
Catalog # 60004-230
Supplier:  Kemtech America
Supplier Number:  H200003
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    Synthware heat transfer block for round bottom flasks, microscale
  • Cat. No.:
    60004-230
  • Supplier no.:
    H200003
Specifications
About this item
Aluminum heat transfer block, used in place of a glass sand bath in microscale experiments.
  • Aluminum heat transfer block
  • Hold 3 flask sizes
Block holds 5 ml, 10 ml, and 25 ml round bottom flasks, with one hole for a standard thermometer.