ContactUsLinkComponent
Fargo® Multi Color Thermal Resin Printer Ribbon
Fargo® Multi Color Thermal Resin Printer Ribbon
Catalog # 77531-626
New Product
Fargo® Multi Color Thermal Resin Printer Ribbon
Catalog # 77531-626
Supplier Number:  SRX45010
New Product
Specifications
  • Color:
    Black, cyan, magenta, yellow
  • Description:
    Multicolor thermal resin printer ribbon, 8.13×10.40×6.02" (W×H×D), 2.875 lbs.
  • Environmentally Preferable:
  • Cat. No.:
    77531-626
Specifications
About this item
The Fargo® 45010 YMCKOK color ribbon is a premium quality ribbon for Fargo DTC1000 and DTC1250 plastic card printers. It is a full-color dye with an overlay panel for extra protection. Plus, this ribbon also comes with two sets of black resin dye that enable dual-sided printing and are essential for producing readable barcodes.
  • 200 full-color single-sided prints
  • YMCKOK ribbon (Yellow, Magenta, Cyan, Black, Overlay and Black panels)
  • Includes cleaning roller
Custom dye-sublimation and resin ribbons work seamlessly with HID printers to consistently produce colorful images, crisp text and precise bar codes.