BioRobotix® Pipette Tips for Rosys® and QIAGEN® Workstations, Molecular BioProducts
Supplier: Thermo Scientific
Certificates
About this item
Compatible with Rosys® and QIAGEN® style workstations.
- Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
Available as liquid sensing tips to maximize the workstation abilites, or clear polypropylene for applications that do not require liquid sensing. Three volume sizes include 250, 300 and 1100 µl. Filter tips utilize the ART® barrier.
Ordering information: Tips are packaged 96 per tray, 10 trays per case.
Specifications
- Catalog No:
- 72830-244
- 72830-248
- 72830-252
- 72830-258
- 72830-256
- 72830-262
- 72830-260
- 72830-266
- 72830-264
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental Certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)