Specifications
- Description:Ictotest Urinalysis Reagent Tablets
- Environmentally Preferable:
- Cat. No.:AM2591
- Parameters tested:Bilirubin
- Supplier no.:2591
Specifications
About this item
Ictotest® Reagent Tablets from Siemens Healthineers are used to test for the presence of bilirubin in urine
- Detects bilirubin
- Easy to use
- Results in one minute
The presence of bilirubin is an important finding in the evaluation of liver function and a positive detection could be an indication of liver disease such as jaundice, cirrhosis or hepatitis. Hospital and laboratories rely on the quality of Siemens’ Ictotest reagent tablets to confirm bilirubin results prior to providing the results to their customers. Use of Ictotest requires a dropper and distilled water, in addition to the absorbent mats and reagent tablets provided. The test is easy to perform and gives timely and accurate confirmatory urine bilirubin results in one minute. Level of detection ≥ 0.05 to 0.1 mg bilirubin/dL in urine (0.9 to 1.7 µmol/L). Tablets are packaged 100 per bottle.