Order Entry
Puerto Rico
ContactUsLinkComponent
OSOM® Strep A Test Kit, Sekisui Diagnostics
OSOM® Strep A Test Kit, Sekisui Diagnostics
Catalog # 71001-048
CAS Number:  
undefined
OSOM® Strep A Test Kit, Sekisui Diagnostics
Catalog # 71001-048
Supplier Number:  141
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Strep A Test Kit
  • Cat. No.:
    71001-048
  • Supplier no.:
    141
  • No. of tests:
    50

Specifications

About this item

The OSOM® Strep A test kit provides clear, accurate results in as little as five minutes

  • Sensitivity 96%
  • Specificity 98%

The easy-to-read two-color results reduce patient waiting time and allow for quick diagnosis. Specimens are obtained via throat swab. Test kits can be stored for 17 months at room temperature.

Certifications: CLIA-waived. CE marked.
Ordering information: Kit includes two additional tests for external QC testing (50-test kit includes 52 tests).