Order Entry
Puerto Rico
ContactUsLinkComponent
VDRL Syphilis Test Kit, Arlington Scientific
VDRL Syphilis Test Kit, Arlington Scientific
Catalog # 80087-360
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  
undefined
VDRL Syphilis Test Kit, Arlington Scientific
Catalog # 80087-360
Supplier Number:  950010
Product available on GSA Advantage®
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    VDRL Syphilis Test Kit
  • Cat. No.:
    80087-360
  • Supplier no.:
    950010

Specifications

About this item

The ASI VDRL Antigen test for syphilis is a specific qualitative and semi-quantitative nontreponemal flocculation test for the detection of reagin antibodies in human serum. The procedure is based on the VDRL antigen being combined at a correct ratio with buffered saline and then mixed with heat-inactivated serum.

  • Suitable for testing cerebral spinal fluid (CSF) specimens
  • Meets CDC primary algorithm recommendation
  • Easy to interpret
  • Simple and economical to use
  • Screw cap vials - No glass ampules
  • Room temperature storage
  • Longest shelf life - up to 60 months
  • Proudly made in USA
  • Results on CAP, AAB and API surveys
  • Sensitivity & Specificity
  • Sensitivity: >99%
  • Specificity: >99%
Certifications: CE marked. CPT Code 86592.
Ordering information: Kits include two vials of VDRL antigen (2 x 5 mL), two vials of VDRL buffered saline (2 x 60 mL). The kit could perform as many as 5,900 tests but it is generally less due to facility testing workflow.