Automated Tests, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Reagents must be stored at 2 to 8°C (36 to 46°F)
Reagents for total protein tests can be stored at 2 to 25°C (36 to 77°F).
Reagents must be stored at 2 to 8°C (36 to 46°F)
Reagents for total protein tests can be stored at 2 to 25°C (36 to 77°F).