Order Entry
Puerto Rico
ContactUsLinkComponent
Pierce ProFound™ Biotinylated-Protein Interaction Pull Down Kit, Thermo Scientific
Pierce ProFound™ Biotinylated-Protein Interaction Pull Down Kit, Thermo Scientific
Catalog # PI21115
Supplier:  Invitrogen
CAS Number:  
undefined
Pierce ProFound™ Biotinylated-Protein Interaction Pull Down Kit, Thermo Scientific
Catalog # PI21115
Supplier:  Invitrogen
Supplier Number:  21115
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Biotinylated-Protein:Protein Interaction Kit
  • Cat. No.:
    PI21115
  • No. of reactions:
    25 Pull-down reactions, each using 25 μl resin
  • Supplier no.:
    21115

Specifications

About this item

Pierce™ Biotinylated Protein Interaction Pull-Down Kit contains the necessary components to capture and purify interactors of a biotin-labeled protein or ligand.

  • Provides a complete easy-to-use strategy for discovering protein:protein interactions
  • Uses common laboratory equipment and reagents
  • Adaptable to single- or multiple-sample demands
  • Flexible pull-down format uses spin cups for easy and efficient manipulation of streptavidin agarose beads
Delivery: Kit includes streptavidin Agarose resin (1.5 mL) settled gel; Lysis Buffer (250 mL); BupH Tris Buffered Saline, 1 pack (500 mL); Wash Buffer (Acetate, pH 5), (100 mL); Elution Buffer (pH 2.8), (50 mL); Spin Columns Accessory Pack, 27 columns; Collection tubes and caps (200x 2 mL tubes).