Orders LinkContactUsLinkComponent
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
These 96-well microplates are available with flat-, round-, conical-, and C-bottom well styles, and are manufactured from clear, virgin polystyrene and selected for optical clarity and consistency.
  • Clear virgin polystyrene construction
  • High degree of optical clarity
  • Chimney-well design helps reduce cross-contamination
  • Designed to minimize air bubble formation
C-bottom plates feature a flat bottom for optical measurement, but the hard corners are curved to eliminate liquid traps and improve plate washing
All plates are free of detectable RNase, DNase, human DNA, and pyrogens.
MICROLON™ 200 and MICROLON™ 600 formats are available for consistent performance in binding assays. The chimney-well design option reduces the risk of cross-contamination and facilitates sealing by cap mat, thermal sealer, or adhesive sealer. Plates are supplied without lids.
Product Family Options
MICROLON™ 200 Medium Binding 96-Well Microplates
  • Color
    Surface treatment
    Well shape
  • Clear
    Medium binding
    Conical-Bottom
    Case of 40
    Case of 40 - $146.00
    $146.00
    Case of 40
       $146.00
     : 651001
  • Clear
    Medium binding
    Flat-Bottom
    Case of 40
    Case of 40 - $123.00
    $123.00
    Case of 40
       $123.00
     : 655001
  • Clear
    Medium binding
    Flat-Bottom, with Chimney
    Case of 40
    Case of 40 - $123.00
    $123.00
    Case of 40
       $123.00
     : 655080
  • Clear
    Medium binding
    Round-Bottom
    Case of 40
    Case of 40 - $107.00
    $107.00
    Case of 40
       $107.00
     : 650001
MICROLON™ 600 High Binding 96-Well Microplates
  • Color
    Surface treatment
    Well shape
  • Clear
    High binding
    Conical-Bottom
    Case of 40
    Case of 40 - $148.00
    $148.00
    Case of 40
       $148.00
     : 651061
  • Clear
    High binding
    C-Bottom, with Chimney
    Case of 40
    Case of 40 - $157.95
    $157.95
    Case of 40
       $157.95
     : 655085
  • Clear
    High binding
    Flat-Bottom
    Case of 40
    Case of 40 - $123.00
    $123.00
    Case of 40
       $123.00
     : 655061
  • Clear
    High binding
    Flat-Bottom
    Case of 150
    Case of 150 - $569.32
    $569.32
    Case of 150
       $569.32
     : 655061-25
  • Clear
    High binding
    Flat-Bottom, with Chimney
    Case of 40
    Case of 40 - $120.00
    $120.00
    Case of 40
       $120.00
     : 655081
  • Clear
    High binding
    Round-Bottom
    Case of 40
    Case of 40 - $113.00
    $113.00
    Case of 40
       $113.00
     : 650061
Standard 96-Well Microplates
  • Color
    Surface treatment
    Well shape
  • Clear
    Untreated
    Conical-Bottom
    Case of 100
    Case of 100 - $129.00
    $129.00
    Case of 100
       $129.00
     : 651101
  • Clear
    Untreated
    Flat-Bottom
    Case of 100
    Case of 100 - $140.00
    $140.00
    Case of 100
       $140.00
     : 655101
  • Clear
    Untreated
    Round-Bottom
    Case of 100
    Case of 100 - $137.00
    $137.00
    Case of 100
       $137.00
     : 650101