ContactUsLinkComponent
MIC 2000™ 96-Well Plate, Thermo Scientific
MIC 2000™ 96-Well Plate, Thermo Scientific
  62403-051
undefined
MIC 2000™ 96-Well Plate, Thermo Scientific
  62403-051
 :  12-0151
  • Well volume:
    0.28 mL
  • Cat. No.:
    62403-051
  • Supplier no.:
    12-0151
The MIC 2000™ is a Minimum Inhibitory Concentration plate
It is a 96-well plate with U-bottom wells. Plate is ideal for microbiological applications using a Microtiter® format. For use with any multichannel pipettor or liquid handling instrument with standard 9mm spacing. Compatible with Thermo Scientific universal microplate lids (see 62402-929 series). Sterile.