About this item
Kits include all the components needed for specimen collection, smear preparation, fixation, and courier transport to the laboratory
They provide excellent structural support for the specimen slide and adhesive-free slide attachment allowing it to be easily removed from the folder. Kits contain various combinations of one or two slides, cervex brushes, wood scrapers, cytology brushes, cotton swabs, and fixative sprays and pouches. Cervex brushes collect all essential cells for tracing the early stages of cervical cancer. The bristles collect endo-, ectocervical, and T-zone cells. The soft, flexible bristles reduce the risk of injury during examination. Cytology brushes allow for collection of endocervical, Chlamydia, and HPV cells, and reduce the chances of false negative specimens. Sprays and pouches contain a non-aerosol fixative made of a water and alcohol soluble polyethylene glycol formulation used for affixing clinical specimens. A tough thin film forms to maximize cell preservation and ensure an optimum staining environment. Fixative is available in spray pump bottles or single-use foil pouch pack.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Related Information
Agilent Molecular and Syn Bio solutions
Product Literature
VWR GHS Compliance Letter (US)