Nunc® CovaLink™ 96-Well Module, Thermo Scientific
Catalog # 62409-285
Supplier: Thermo Fisher Scientific
Specifications
- Well volume:400 µL
- Color:Clear
- Cat. No.:62409-285
- Well shape:flat-bottom
- Supplier no.:478042
Specifications
About this item
CovaLink™ module is made of optically clear polystyrene for covalent binding of biomolecules
Module consists of 96 wells (8 wells/strip, 12 strips/frame) within a one-way frame for precise lid orientation.
The 96-well format allows use of standard laboratory equipment and existing assay protocols. CovaLink™ module provides improved stability and minimal steric inhibition via a specialized binding surface containing secondary amino groups, which are tethered to the end of a spacer arm. Biomolecules are efficiently coupled using common chemistries such as carbodimide.