Nunclon™ Delta 96-Well MicroWell™ Optical Bottom Plates, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
These polystyrene plates provide optimum clarity for viewing well contents
The Nunclon™Δ treated surface ensures optimal cell growth and attachment. Plates feature either a black or white polystyrene upper structure and a polymer bottom. The flat-bottom well design facilitates plate reader access. The 96-well footprint is compatible with standard equipment and automated systems.
Ordering information: Both plates come supplied with lids. For sealing tapes, see the 62409-118 series.
Related Information
Certificates
Search by Lot NumberThermo Scientific Nunc Cell Culture Plastics Guide