Order Entry
Puerto Rico
ContactUsLinkComponent
 
BTXpress™ Cytoporation® Electroporation Buffer, Medium T and T4
Catalog # 77552-850
Supplier:  Harvard Apparatus
New Product
CAS Number:  
undefined
BTXpress™ Cytoporation® Electroporation Buffer, Medium T and T4
Catalog # 77552-850
Supplier:  Harvard Apparatus
Supplier Number:  47-0002
New Product
CAS Number:  

Specifications

  • Description:
    Electroporation buffer, BTXpress Cytoporation® Medium T
  • Size:
    500 ml
  • Cat. No.:
    77552-850

Specifications

About this item

BTXpress Cytoporation® Media T/T4 is an advanced electroporation buffer designed for use with the BTX AgilePulse MAX™ large volume electroporation system for in vitro delivery of DNA, RNA, oligonucleotides, and siRNA. The low conductivity buffer is specially formulated to minimise heating of solution during large volume electroporation for maximum transfection efficiency and high cell viability. BTXpress Cytoporation® Media T and T4 are sterile filtered from the highest quality non animal, medical grade reagents. Buffer can be directly diluted in complete growth media for post electroporation cell culture.

  • Optimised for eukaryotic electroporation applications
  • Maximum transfection efficiency, high cell viability
  • Low conductivity means minimal heating or electrode arcing
  • Physiological pH and balanced osmolarity
  • Contains no animal products
  • Certified free from RNase and DNase

Two formulations with different conductivities are available for optimum conductivity for each eukaryotic cell type.