Adherent Cell Electrode
Product Details & Documents
Reusable plate electrode designed to directly electroporate adherent cells or electroporate cultured cell monolayers on filters. The adherent cell electrode is a reusable electroporation applicator designed to fit into each single well of a 24-well or 12-well plate.
This plate electrode is specially designed to electroporate cultured monolayers of cells on laminin-coated filters, or on 12 mm polycarbonate or polyester transwell filter units (0.4 mm pore size). It can directly electroporate adherent cells such as human corneal keratocytes, Caco-2, dental pulp cells, NC65 and BCEC and MDCK cells.
The adherent cell electrode consists of two parallel plate electrodes, 7×19 mm with 5 mm gap spacing between the plates. Each plate electrode has a 0.3 mm height insulated foot at both ends of the bottom. When the electrode is placed in a dish, a foot minimises the damage on cells. The high voltage cables are connected to a BTX electroporator, and the electrode is brought down onto the filter until it makes contact with the buffer, usually at 1 to 2 mm above the filter. A combination of HV and LV pulses may be used. Adherent cell electrodes are compatible with Gemini X2, ECM 830, ECM 2001+, and Legacy ECM 2001 generators.
Specifications
- Description:Adherent cell electrode, 5 mm gap
- Cat. No.:77552-848
- Pulse Length Range:1 µs - 35 ms
- Voltage Range:0 - 300 V
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



