Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Leptin ELISA

Leptin ELISA

Leptin ELISA, 1×96 tests
Catalog # 101720-304
Marketsource
Supplier:   Alpco Diagnostics
Supplier #:  11-LEPHU-E01
Leptin ELISA
Catalog # 101720-304
Supplier:  Alpco Diagnostics
Marketsource
CAS Number:  
Leptin ELISA

Leptin ELISA, 1×96 tests

CAS Number:  
Price
$600.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Leptin ELISA is for the quantitative determination of leptin in human serum.

  • Analytical sensitivity 0.42 ng/ml
  • Dynamic range 1 to 100 ng/ml

The Leptin ELISA follows a typical two-step sandwich type assay format. The Leptin ELISA makes use of two highly specific monoclonal antibodies: A monoclonal antibody specific for leptin is immobilized onto the microWells plate and another monoclonal antibody specific for a different epitope of leptin is conjugated to biotin. The Leptin ELISA includes all components (96 well Ab coated plate, conjugate Ab, substrate, detector, standards, quality controls and dilution and wash buffer) necessary to complete the Leptin ELISA.

 : Research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    Leptin ELISA
  • Size:
    1×96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    1 - 100 ng/ml
  • Time to Results:
    1 h, 45 min
  • Detection Method:
    Colorimetric
  • Cat. No.:
    101720-304
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Sensitivity:
    0.42 ng/ml
  • Conjugate ELISA:
    Biotin
  • Format:
    Pre-coated
  • Assay Type (ELISA with LOV):
    Sandwich
  • Target protein:
    Leptin
  • Sample Type:
    serum

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.