Order Entry
Puerto Rico
ContactUsLinkComponent
Ultrasensitive Insulin ELISA Jumbo (10-plate)
Ultrasensitive Insulin ELISA Jumbo (10-plate)
Catalog # 102977-826
Supplier:  Alpco Diagnostics
CAS Number:  
Ultrasensitive Insulin ELISA Jumbo (10-plate)
Catalog # 102977-826
Supplier:  Alpco Diagnostics
Supplier Number:  80-INSHUU-E10
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type (ELISA with LOV):
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Insulin
  • Description:
    Ultrasensitive insulin ELISA jumbo (10-plate)
  • Size:
    10×96 tests
  • Sample Type:
    Plasma, Serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h, 30 min
  • Detection Range:
    0.15 - 20 µIU/ml
  • Sensitivity:
    0.135 µIU/ml
  • Regulatory Status:
    FDA Class I device. For in vitro diagnostic use
  • Cat. No.:
    102977-826
  • Supplier no.:
    80-INSHUU-E10

Specifications

About this item

The ultrasensitive insulin ELISA jumbo (10-plate) kit quantifies the concentration of insulin protein in human serum or plasma samples.

  • Only 25 µl of sample is sufficient for the assay
  • Superior sensitivity of 0.135 µIU/ml
  • Dynamic range of 0.15 to 20 µIU/ml

The ultrasensitive insulin ELISA kit is a FDA registered in vitro diagnostic tool for the quantification of human insulin in a clinical setting or research laboratory. The Ultrasensitive Insulin ELISA utilizes a dual-monoclonal antibody sandwich ELISA format, diabetes control, and 96-well microplate comprising removable strips. The Ultrasensitive Insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if the entire kit is not needed at one time.

Certifications: FDA Class I device. For in vitro diagnostic use.
Packaging: Single kit packaging is also available.