Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay duration:Multiple steps
- Assay Type (ELISA with LOV):Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Human
- Target protein:Insulin
- Description:Insulin ELISA jumbo (10-Plate)
- Size:10×96 tests
- Sample Type:Plasma, Serum
- Detection Method:Colorimetric
- Time to Results:1 h, 15 min
- Detection Range:3 - 200 µIU/ml
- Sensitivity:0.399 µIU/ml
- Regulatory Status:FDA Class I device. For in vitro diagnostic use
- Cat. No.:101721-582
- Supplier no.:80-INSHU-E10.1
The insulin ELISA jumbo (10-plate) quantifies the concentration of insulin protein in human serum or plasma samples.
- Only 25 µl of sample is sufficient for the assay
- Superior sensitivity of 0.135 µIU/ml
- Dynamic range of 0.15 to 20 µIU/ml
The insulin ELISA jumbo (10-plate) is a FDA registered in vitro diagnostic tool for the quantification of human insulin in a clinical setting or research laboratory. Utilizing a dual-monoclonal antibody sandwich ELISA format, bi-level control set, and 96-well microplate comprising removable strips, the insulin ELISA kit has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if an entire plate is not needed at one time.