DuPont™ Tyvek® 400 SFR garments are lightweight, disposable secondary flame-resistant overgarments designed to help protect and preserve primary flame-resistant garments, while providing an ideal balance of protection, durability, and comfort.
- Red trim elements - red external stitched seams, red zipper, and red chest patch border
- Patent-pending design
- Laydown collar
- Storm flap
- Open wrists/open ankles
- Seam: Serged (external), red
Relaxed fit coverall design features a diamond gusset crotch to allow for unrestricted movement and optimal fit when wearing primary flame-resistant protective clothing beneath.
Composed of flash spun high density polyethylene breathable nonwoven Tyvek® fabric that has been flame-retardant treated, available only from DuPont, Tyvek® 400 SFR garments offer an inherent barrier against particles (down to 1.0 micron in size). Applications include: Oil and gas, general manufacturing/operations, maintenance, dirty jobs, environmental services, utilities.
Tyvek® 400 SFR represents a new generation of disposable secondary flame-resistant garment technology. Compared to traditional disposable secondary flame-resistant garments that have been available for years, Tyvek® 400 SFR is a differentiated multi-hazard garment solution, offering superior particle barrier protection and improved durability in a lighter-weight fabric. This unique combination of performance allows Tyvek® 400 SFR garments to be worn over primary flame-resistant garments like Nomex® when hazardous particle and flash fire hazards exist. Additionally, Tyvek® 400 SFR is a more sustainable garment solution and can be recycled through the Tyvek® protective apparel recycling program if not contaminated with hazardous materials.
- Catalog No:
- 77532-352
- 77532-342
- 77532-338
- 77532-362
- 77532-350
- 77532-360
- 77532-366
- 77532-334
- Garment application:
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory
- Industrial, Chemical protection, Laboratory