Order Entry
Puerto Rico
ContactUsLinkComponent
Irradiated String Mop, MegaSorb, Micronova
Irradiated String Mop, MegaSorb, Micronova
Catalog # 76837-146
Supplier:  Micronova
Irradiated String Mop, MegaSorb, Micronova
Catalog # 76837-146
Supplier:  Micronova
Supplier Number:  MZCRMS11-12IR

Specifications

  • Description:
    MegaSorb microfiber string mop, irradiated
  • Cat. No.:
    76837-146
  • Width:
    15.2 cm (6 ")

Specifications

About this item

A microfiber string mop for use with the QDMH-1 or MH-1 series mop adapters. Designed for cleanrooms and controlled environments.

Made from 70% polyester/30% nylon.

Packaging: Single bagged. Irradiation indicator placed on outer bag. 12 mops are then packed into a polylined case. Irradiation indicator placed on case.