2-Bromocyclopentanone ≥95%
Caution: for Research Use Only
- :21943-50-0
- :C₅H₇BrO
- :MFCD11109944
- :163.01 g/mol
- :1760
- :8,Ⅲ
Caution: for Research Use Only
- :21943-50-0
- :C₅H₇BrO
- :MFCD11109944
- :163.01 g/mol
- :1760
- :8,Ⅲ
Product Family Options
- Pack typeSize
- : A826077-100MGNew Product
Specifications
Cat. No.78531-786New Product - : A826077-250MGNew Product
Specifications
Cat. No.77901-918New Product - : A826077-1GNew Product
Specifications
Cat. No.77900-520New Product - : A826077-5GNew Product
Specifications
Cat. No.77900-913New Product - : A826077-10GNew Product
Specifications
Cat. No.77901-452New Product - : A826077-25GNew Product
Specifications
Cat. No.77901-149New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



