Order Entry
Puerto Rico
ContactUsLinkComponent
Propyl hexanoate ≥98%
Flammable
Irritant
Propyl hexanoate ≥98%
Catalog # 77602-574
Supplier:  Ambeed
New Product
CAS Number:  
Flammable
Irritant
Propyl hexanoate ≥98%
Catalog # 77602-574
Supplier:  Ambeed
Supplier Number:  A623455-25G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    25 g
  • Cat. No.:
    77602-574

Specifications

About this item

 :
 :158.24 g/mol
UN:3272
ADR:3,Ⅲ
 :GHS02
 :GHS07
CAS Number:626-77-7
Formula:C₉H₁₈O₂
MDL Number:MFCD00053803