AqueLyn 7 PLUS Neutral Cleaner, Quip Laboratories
Catalog # 10121-156
Supplier: Quip Laboratories
Specifications
- Description:AqueLyn 7 PLUS Neutral Cleaner
- Size:113,5 L (30 gal)
- Cat. No.:10121-156
- Packaging:Drum
- Supplier no.:AQ7PL-30GAL
Specifications
About this item
AqueLyn 7PLUS is an industry-proven, high-performance neutral cleaner designed to effectively clean a variety of applications from cage and rack washing, to windows, mirrors, walls, and restrooms.
- Non-hazardous and safe for manual cleaning of equipment
- Biodegradable
- Excellent for everyday use from light-duty cleaning to heavy-duty cleaning
- Highly effective for daily, damp mopping
- Improved tolerance for cleaning with hard water