Order Entry
Puerto Rico
ContactUsLinkComponent
Roto-Rock Genie Rotator Rocker, 120 V
Roto-Rock Genie Rotator Rocker, 120 V
  77574-960
New Product
undefined
Roto-Rock Genie Rotator Rocker, 120 V
  77574-960
 :  SI-R100
New Product

 

  • Rotation Speed (min-1):
    1 - 70 cycles/min; 1 - 35 rpm
  • Temperature:
    0...38 °C
  • Description:
    Roto-Rock Genie, type B (US/CA), 120 V
  • Electrical:
    Input: 90 - 264 V AC, 50/60 Hz
  • Shipping Weight:
    10.2 kg (22.5 lbs.)
  • Max. Load:
    4.5 kg (10 lbs.)
  • Platform Size:
    320×250 mm
  • Dimensions:
    400×300×380 mm
  • Timer:
    1 s - 99 h, 59 min, 59 s, or continous
  • Weight:
    9.7 kg (21.3 lbs.)
  • Rocking Angle:
    ±1 - 25° from horizontal
  • Cat. No.:
    77574-960

 

 

Programmable combination rotator rocker with digital touch screen, featuring magnetic platform and clip plate system.

  • 120 V model
  • Intuitive touch screen control makes changing program settings easy
  • Setting the tilt angle and speed control provides thorough mixing while eliminating spills when using open containers in rocking mode
  • The two-sided magnetic platform holds virtually any type of tube and other sample vessels used in the laboratory when using the GENIE clip plates (sold separately)
  • Compact design saves valuable bench space in the laboratory
  • Designed for use in cold rooms, incubators (non-CO₂), or at ambient temperature
  • 2 year warranty

Programable all-in-one benchtop GENIE Rocker/Rotator or cycle from Rock to Rotate. Digital set/control for rpm, total time, cycle time. Features 'pause/resume' button allowing the operator to add reagents to a sample during processing.

Programming modes: Rock, rotate, or timed auto-switching between rock and rotate
Rocking tray size: 300×300 mm
Power: DC 24 V, 2.5 A

 : CE certified.
 : Supplied with magnetic platform and non-slip rocking tray. Accessories must be ordered separately: GENIE clip plate systems designed to handle a variety of vessels from PCR tube, vials, bottles and sealed bags.