OneTouch™ Filter Barrier Pipette Tips, Sterile, Sorenson BioScience
Supplier: Sorenson Bioscience
Certificates
About this item
These pipette tips combine virgin polypropylene and pliable TPE to provide a rigid and dependable tip with a soft.
- Dual-shot tip design
- Easy-to-seal
- Color-coded collar
The innovative design creates an ergonomic tip that can be used across a wide variety of pipettors.
Packaging: Available in traditional plastic racks, or edek™ paperboard racks that fit seamlessly into paper recycling programs.