ContactUsLinkComponent
Nunc-Immuno™ TSP Replicator, MaxiSorp™, Thermo Scientific
Nunc-Immuno™ TSP Replicator, MaxiSorp™, Thermo Scientific
Catalog # 62409-179
undefined
Nunc-Immuno™ TSP Replicator, MaxiSorp™, Thermo Scientific
Catalog # 62409-179
Supplier Number:  445497
Specifications
  • Description:
    Nunc-Immuno™ TSP Replicator, MaxiSorp™
  • Cat. No.:
    62409-179
  • Supplier no.:
    445497
Specifications
About this item
This Transferable Solid Phase (TSP) disposable replicator is similar to a 96-well plate lid with 96 attached pins.
  • Suitable for the adsorption of mixed proteins or antibodies directly from cultures
  • Sterilized, polystyrene
When precoated with antigen or antibodies, a specific analyte from a culture may be assayed, via efficient transfer from one plate to another. Can be assayed in the Nunc MicroWell™ 96-well plates.
When placed over the well plate, the pins are immersed in the wells. MaxiSorp™ is ideal for replating and screening of 96-well plates containing transformants, library clones, screening hybridoma cultures, and expression libraries.