VWR® Contour™ End Upright Support
End upright support, Light neutral, Steel, 18½", 12", 2"
Catalog # CFW-0014-1A
Supplier: VWR International
Jump to:
Product Details & Documents
The end unit supports the end of the reagent rack assembly where facia panels are used.
- Color: Light neutral
Specifications
- Width:18½"
- Color:Light neutral
- Description:End upright support
- Height:2"
- Length:12"
- Material:Steel
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



