ContactUsLinkComponent
BioCoat™ HTS Caco-2 Assay Systems, PET Membrane, Corning
BioCoat™ HTS Caco-2 Assay Systems, PET Membrane, Corning
Catalog # 12777-042
Supplier:  Corning
   Low Manufacturing Impact
undefined
BioCoat™ HTS Caco-2 Assay Systems, PET Membrane, Corning
Catalog # 12777-042
Supplier:  Corning
Supplier Number:  354802
   Low Manufacturing Impact
Specifications
  • Description:
    HTS Caco-2 assay system, inc. media and supplements, for 120 individual 3-day Caco-2 assays
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO 14001 - Bedford (reg. no. 14.4942)
  • Cat. No.:
    12777-042
  • Supplier no.:
    354802
Specifications
About this item
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 assay system, PET membrane contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat fibrillar collagen 24-well insert system.