ContactUsLinkComponent
Infecon® 2000 Infectious Substance/Diagnostic Specimen Shipper, Com-Pac
Infecon® 2000 Infectious Substance/Diagnostic Specimen Shipper, Com-Pac
Catalog # 11217-570
Supplier:  Com-Pac
undefined
Infecon® 2000 Infectious Substance/Diagnostic Specimen Shipper, Com-Pac
Catalog # 11217-570
Supplier:  Com-Pac
Supplier Number:  INF-2000
Specifications
  • Dimensions:
    10.2L×10.2W×20.3H cm (4×4×8")
  • Cat. No.:
    11217-570
  • Supplier no.:
    INF-2000
Specifications
About this item
Economical, dual-purpose shipper is ideal for shipping test tubes containing infectious substances or diagnostic specimens.
The durable outer box is clearly marked with UN certification markings.
Shipper complies with UN regulations, ICAO/IATA Packing Instruction 602, and applicable DOT regulations.
Ordering information: Includes a 50mL (1.7oz.) capacity absorbent sheet, a liquid-tight, 500mL (16.9oz.) pressure vessel that is able to withstand up to 95kPa of pressure, and a compartmental bubble pouch to separate test tubes and provide cushioning support. Shipper’s declarations and packing instructions are included. Also comes with an infectious substance label and a diagnostic specimen label. Dimensions measurements include 500mL vessel.