ContactUsLinkComponent
Dry Ice Infectious Substance Shipper, Qorpak®
Dry Ice Infectious Substance Shipper, Qorpak®
Catalog # 16455-262
Supplier:  Berlin Packaging
Small Business Enterprise
undefined
Dry Ice Infectious Substance Shipper, Qorpak®
Catalog # 16455-262
Supplier:  Berlin Packaging
Supplier Number:  HMS-6905M
Small Business Enterprise
Specifications
  • Description:
    Dry ice infectious substance shipper
  • Environmentally Preferable:
  • Cat. No.:
    16455-262
  • Supplier no.:
    HMS-6905M
Specifications
About this item
Certified packaging for shipment of infectious substances.
  • Acceptable for ground and air shipment
  • Will accept up to 12 vials (vials not included)
Certified for Class 6.2 Infectious Substances.
Styrofoam core mount is suspended to accommodate dry ice. Complete with corrugated outer carton, die-cut foam, absorbent material, metal sealing ring, plastic bag, and tape. Warning labels included.