Ethanol ≥70%
Plastic bottle, 250 ml
Catalog # 77540-210
Supplier: G-Biosciences
Supplier #: 786-2076
Jump to:
Product Details & Documents
Use for differential staining of Bacteria.
- An aqueous 70% solution
Quick sterilizing solution
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
Specifications
Specifications
- Pack type:Plastic bottle
- Size:250 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



