ContactUsLinkComponent
OIML Class F1 Standard Weight Set
OIML Class F1 Standard Weight Set
Catalog # 11238-102
Supplier:  Mettler Toledo
undefined
OIML Class F1 Standard Weight Set
Catalog # 11238-102
Supplier:  Mettler Toledo
Supplier Number:  30406408
Specifications
  • Description:
    F1 OIML class standard weight set (plastic case)
  • Cat. No.:
    11238-102
  • Weight:
    1 - 500 mg
  • Includes:
    12 pieces, with certificate
  • Supplier no.:
    11119581
Specifications
About this item
Weight calibration performed by robots: State-of-the-art robot technology minimizes errors and increases accuracy.
  • Stainless steel
  • Density: 7950 (±140) kg/m³
Each knob weight has an adjusting cavity, which simplifies weight adjustment without the need for specialized equipment.
Certifications: Calibration certificate in accordance with ISO/IEC 17025 requirements.