InLab® Redox Ag Combination Electrode, METTLER TOLEDO®
97001-690
Product available on GSA Advantage®
- Temperature range:0...100 °C
- Length:120 mm
- Material:Glass
- Cat. No.:97001-690
- Internal reference:ARGENTHAL™
- Junction:Ceramic
- Connector:S7
- Supplier no.:51343205
- Electrolyte:1M KNO3
Electrode measures oxidation/reduction power of a solution
- Silver ring indicator
- Safelock™ closing ring
- ARGENTHAL™ lead-off and silver ion trap
- S7 screw head connector
Features a silver ring indicator, a Safelock™ closing ring for easy refilling and complete sealing, and an ARGENTHAL™ lead-off and silver ion trap for optimum silver ion-free accuracy at various sample temperatures. S7 screw head connector is compatible with various cables, so electrode may be used with a wide range of meters.