Orders LinkContactUsLinkComponent
Disposable Face Shields
Disposable Face Shields
Supplier:   Hardy Diagnostics
Disposable Face Shields
Supplier:  Hardy Diagnostics
Certificates
Disposable Face Shields
Disposable Face Shields
1 Options Available Below
Product Details & Documents
Face shields are personal protective equipment (PPE) that are used for the protection of the facial areas (eyes, nose, mouth) from sprays, aerosols, and splashes.
  • Latex free
  • Made in an FDA registered facility
  • Made from PETG
  • Provides clarity of view
  • Visors are anti-fogging
  • Once size fits most
  • Includes a comfortable forehead cushion
  • Protective film on the shield (must be removed before use)
Ideal for healthcare and laboratory workers. Well suited for protection against COVID-19 and other pathogens.
Industries: Breweries, wineries and distilleries, Cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing.
Documents
Product Family Options
Product Information
  • Color
    Material
    Availability
    Price
  • 76423-492
    Face shield, full face, disposable, anti-fogging plastic
    Face shield, full face, disposable, anti-fogging plastic
    Clear
    PETG
    Log in for availability
    Log in for availability
    Box of 24
    Box of 24 - $128.00
    $128.00
    Box of 24
       $128.00
    Supplier Number: 4511
  • 76423-496
    Face shield, full face, disposable, anti-fogging plastic
    Face shield, full face, disposable, anti-fogging plastic
    Clear
    PETG
    Discontinued
    Supplier Number: MD30331