OPTICHEM™ Blocks for Centrifugal Vacuum Evaporators, Chemglass
Supplier: Chemglass
Certificates
About this item
Vial and tube holders are for use in centrifugal vacuum evaporators.
Lightweight blocks maximize evaporation without increasing sample volume. Black hardcoat anodized aluminum construction improves resistance against organic solvents. Blocks are laser-etched with vial and tube sizes.