Order Entry
Puerto Rico
ContactUsLinkComponent
 
Safe-Lock Microcentrifuge Tubes, Polypropylene, Eppendorf®
Catalog # 89202-684
Supplier:  Avantor
undefined
Safe-Lock Microcentrifuge Tubes, Polypropylene, Eppendorf®
Catalog # 89202-684
Supplier:  Avantor

Specifications

  • Color:
    Clear
  • Cat. No.:
    89202-684
  • Max. rcf:
    30000×g
  • Supplier no.:
    022363743

Specifications

About this item

Eppendorf® Safe-Lock Tubes are the result of 50 years of continuous improvement and development. The original Safe-Lock tubes protect samples with excellent centrifugation stability, feature perfect sealing properties, and reliably prevent sample evaporation.

Ideal for high-speed centrifugation, high-temperature incubations, enzyme assays, and cloning procedures.

One-piece, polypropylene Safe-Lock tubes have flat-top writing surfaces for easy sample identification. Sealing cap lock snaps closed and stays closed while centrifuging, yet opens easily with one hand. Autoclavable at 121 °C (250 °F). Convenient color selection aids in sample identification.

Ordering information: Assorted color option includes natural, blue, green, red and yellow.